Learn More
Waters Corp Waters XTerra MS C18 Sentry Guard Cartridge, 5 μm, 125 Å, 2.1 mm x 20 mm
Supplier:  Waters Corp 186000652
Encompass_Preferred
XTerra MS C18 columns are general purpose, hybrid-based, reversed-phase C18 columns designed for compatibility with mass spectrometry applications. These first-generation columns offer superior pH stability compared to silica-based columns.
  • Hybrid-based
  • Reversed-phase C18
  • Compatible with mass spectrometry applications
  • Superior pH stability compared to silica-based columns
Catalog No. 50-818-786