Learn More
TARGETMOL CHEMICALS INC XU1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T291705MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. XU1(Benzo[c][18]naphthyridin-6(5h)-One) is an Aurora protein kinase inhibitor used for the treatment of diseases suitable for inhibition regulation or modulation of kinase signaling. purity: 99%
Catalog No. 50-258-0460