Learn More
Medchemexpress LLC XVA143 | 264275-77-6 | 905576 | 99.9% | 562.36 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY139202
Encompass_Preferred
XVA143 is an α/β I-like allosteric antagonist that inhibits LFA-1 dependent firm adhesion while simultaneously enhancing adhesion in shear flow and rolling both in vitro and in vivo. It is intended for research use only.
  • Inhibits LFA-1 dependent firm adhesion
  • Enhances adhesion in shear flow and rolling
  • Completely abolishes binding of ICAM-1 to LFA-1
  • Restores rolling of αL-E310A β2 mutant
Catalog No. 50-003-49092