Learn More
Medchemexpress LLC XVA143 | 264275-77-6 | 99.9% | 562.36 g/mol | C25H21Cl2N3O8 | 5 MG

Supplier: Medchemexpress LLC HY1392025MG
XVA143 is an α/β I-like allosteric antagonist of leukocyte function-associated antigen-1 (LFA-1) used as a research tool to modulate cell adhesion. It inhibits LFA-1-dependent firm adhesion while enhancing adhesion under shear flow and rolling in vitro and in vivo, and is supplied as a high-purity compound for biochemical and cell-based assays.
- High purity suitable for research applications.
- Molecular weight 562.36 g/mol.
- Chemical formula C25H21Cl2N3O8.
- CAS number 264275-77-6 for unambiguous identification.
- Soluble in DMSO at up to 100 mg/mL (requires sonication).
- Recommended storage: powder at -20°C or 4°C; in solution at -80°C or -20°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.