Learn More
Selleck Chemical LLC XYL-1 E4719-1G
SDP
Supplier:  Selleck Chemical LLC E47191G
Encompass_Preferred
XYL-1 is a highly potent inhibitor of PARP7 with and IC50 of 0 6 nM It could enhance the type- interferon signaling in vitro and can be a potential candidate for developing cancer immunotherapy agents
Catalog No. 50-000-41342