Learn More
Selleck Chemical LLC Xylene Cyanol FF E2996-1G
SDP
Supplier:  Selleck Chemical LLC E29961G
Encompass_Preferred
Xylene Cyanol FF is an acid triphenylmethane dye It can be used for histochemical staining of hemoglobin peroxidase or as a tracking dye for DNA sequencing in electrophoresis
Catalog No. 50-000-40626