Learn More
GA International XyliFIL™ – Xylene & Chemical-Resistant Labels for Histology0.9375″ x 0.9375″ / 23.8mm x 23.8mm
Supplier:  GA International XFLA67C11WH
Encompass_Preferred
XyliFIL™ labels are specially designed for histology (microscope slides) applications that require resistance to various chemicals and conditions associated with common histology preparation, such as xylene, ethanol series, xylene substitutes, histology stains, and high temperature (70°C) treatments.Its permanent adhesive ensures the labels remain affixed, even in the harshest environments. Their uniform material design avoids delamination issues and won’t absorb histology stains that can obscure printout and barcodes.Intended for use on flat surfaces, these solvent-resistant labels, along with their printout, can withstand immersion in xylene for up to 5 hours on a microscope slide (thorough testing in end-user application conditions is highly recommended). Resistant as well to long-term exposure and immersion in safer clearing and deparaffinizing xylene substitutes such as EZ Prep, Clearify™, Histo-Clear™, Formula 83™, Pro-Par, and others.
Catalog No. 50-226-8118