Learn More
Apexbio Technology LLC XYLITOL-5G
SDP
Supplier:  Apexbio Technology LLC N17255000
Encompass_Preferred
Xylitol (CAS No 87-99-0) is a sugar alcohol commonly studied in the field of biomedical research derived from natural sources such as fruits and vegetables It functions primarily by interfering with bacterial growth notably inhibiting the adhesion of Streptococcus mutans in the oral cavity thereby reducing dental caries Xylitol s mechanism of action involves disrupting metabolic pathways of bacteria which can lead to reduced acid production It exhibits common in vitro activity in the micromolar concentration range Typical applications include its use in cell culture studies investigations of its antibacterial properties in animal models and drug screening processes to evaluate its efficacy as an antimicrobial agent Experimental concentrations typically depend on the specific research design Extensive studies in various model systems have demonstrated its potential in reducing inflammation and improving oral health outcomes
Catalog No. 50-005-67491