Learn More
Medchemexpress LLC Xylometazoline (hydrochloride) | 1218-35-5 | 99.94% | 25 G

Supplier: Medchemexpress LLC 25GHYB0475
Xylometazoline hydrochloride is an α-adrenergic receptor agonist with a Ki ranging from 0.05 to 1.7 μM. It constricts nasal blood vessels and increases nasal airflow, making it suitable for research related to nose stuffiness and runny nose. This compound appears as a white to off-white solid.
- Acts as an α-adrenergic receptor agonist
- Constricts nasal blood vessels
- Increases nasal airflow
- Used in research for nose stuffiness and runny nose
- White to off-white solid appearance
- Purity of 99.94%
- Molecular weight of 280.84
- Chemical formula C16H25ClN2
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.