Learn More
Medchemexpress LLC Xylopine | 517-71-5 | 98.0% | 295.33 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN95345MG
Encompass_Preferred
Xylopine is an aporphine alkaloid with cytotoxic activity on cancer cells. It induces oxidative stress, causes G2/M cell cycle arrest, and triggers apoptosis in cancer cells.
  • Induces oxidative stress
  • Causes G2/M cell cycle arrest
  • Triggers apoptosis in cancer cells
Catalog No. 50-004-26085