Learn More
Medchemexpress LLC Xylose-1-13C | 70849-21-7 | 331104 | 151.12 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0537S2
Encompass_Preferred
Xylose-1-13C is a ¹³C labeled xylose, a stable heavy isotope that can be incorporated into drug molecules as a tracer for quantitation during drug development. It is intended for research use by experienced personnel in appropriately equipped facilities and is not fully validated for medical applications. The chemical, physical, and toxicological properties have not been completely investigated.
  • Acts as a tracer for quantitation during drug development.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs (as an isotope-labeled compound).
  • Suitable for laboratory chemical applications.
  • For research use only.
Catalog No. 50-003-89829