Learn More
Apexbio Technology LLC Xylose 25990-60-7 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B20675.1
Encompass_Preferred
Xylose is a five-carbon monosaccharide widely utilized in biomedical and biochemical research Xylose serves as a metabolic precursor and signaling component in studies of carbohydrate metabolism It is commonly employed in vitro to investigate cellular uptake mechanisms intracellular sugar metabolic pathways and regulation of gene expression mediated by carbohydrate-inducible promoters Experimentally xylose supports the development and analysis of sugar-inducible expression systems microbial metabolic engineering optimization of cell culture medium formulations and exploration of glycosylation pathways Additionally xylose is used as a substrate for metabolic flux analyses and carbohydrate metabolism research in microbial systems
Catalog No. 50-000-93240