Learn More
Medchemexpress LLC Xylotriose | 47592-59-6 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN24691MG
Encompass_Preferred
Xylotriose is a natural xylooligosaccharide that functions as a bifidogenic factor. It acts as an inducer of the xylanase Xyn5 on the cell surface of *Paenibacillus sp. W-61*, enhancing the activity of the xyn5 promoter and promoting xylan utilization by bacteria. This product is for research use only.
  • Natural xylooligosaccharide
  • Acts as a bifidogenic factor
  • Induces xylanase Xyn5 activity
  • Enhances xyn5 promoter activity
  • Promotes xylan utilization by bacteria
Catalog No. 50-004-21813