Learn More
Medchemexpress LLC Xylotriose | 47592-59-6 | 20 MG

Supplier: Medchemexpress LLC HYN246920MG
Xylotriose is a natural xylooligosaccharide that acts as a bifidogenic factor. It induces the xylanase Xyn5 on the cell surface of Paenibacillus sp. W-61, enhancing the activity of the xyn5 promoter and promoting the utilization of xylan by bacteria.
- Natural xylooligosaccharide
- Acts as a bifidogenic factor
- Induces xylanase Xyn5 on Paenibacillus sp. W-61
- Enhances xyn5 promoter activity
- Promotes bacterial xylan utilization
- White to off-white solid appearance
- Typical purity of 99.79%
- Molecular weight of 414.36
- Chemical formula C15H26O13
- Initially sourced from plants, gramineae, zea mays L.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.