Learn More
Medchemexpress LLC Xylotriose | 47592-59-6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN24695MG
Encompass_Preferred
Xylotriose is a natural xylooligosaccharide that acts as a bifidogenic factor. It functions as an inducer of the xylanase Xyn5 on the cell surface of Paenibacillus sp. W-61, playing a role in bacterial xylan utilization.
  • Oligosaccharide that acts as an inducer of the xylanase Xyn5 on the cell surface of Paenibacillus sp. W-61.
  • Enhances the activity of the xyn5 promoter.
  • Increases the expression of the xyn5 gene.
  • Promotes the utilization of xylan by bacteria.
Catalog No. 50-004-21161