Learn More
Iwai North America Xyltech™ BOF-01: Human pluripotent stem (ES/iPS) cells Proliferation Control Medium. Size: 100mL
Supplier:  Iwai North America B10101
Encompass_Preferred
Xyltech™ BOF-01 is a serum-free novel culture medium that suppresses cell growth while maintaining cell morphology and the ability to differentiate, optimized for human pluripotent stem (ES/iPS) cells. It is perfect for those looking to slow down cell growth to prevent over-confluency.
Catalog No. NC2733034