Learn More
FOCUS BIOMOLECULES Y-27632 dihydrochloride, ROCK family kinase inhibitor, CAS# 129830-38-2, 99% by HPLC_NMR (Conforms)
SDP
Supplier:  FOCUS BIOMOLECULES 1023015MG
Encompass_Preferred
Biochemical Reagents, Y-27632 dihydrochloride, ROCK family kinase inhibitor, CAS# 129830-38-2, 99% by HPLC_NMR (Conforms), Specific inhibitor of ROCK family kinases.1 Enhances stem cell survival and proliferation in culture.2 Significantly improves freeze/thaw survival rate for human embryonic stem cells without influencing morphology, karyotype, cell surface markers, or differentiation potential.3
Catalog No. NC2029263