Learn More
Enzo Life Sciences Y27632 . dihydrochloride, (25mg), CAS Number: 129830-38-2
SDP
Supplier:  Enzo Life Sciences ALX270333M025
Encompass_Preferred
ROCK inhibitor. Y-27632 is a highly potent, cell permeable, selective and ATP competitive inhibitor of ROCK1 and ROCK2 (IC. .There has been a growing interest in Y-27632 for use in stem cell self-renewal and reprogramming. It is known to increase the survival rate of human embryonic stem cells undergoing cryopreservation. Alternative Name: (R)-(+)-trans-N-(4-Pyridyl)-4-(1-aminoethyl)-cyclohexanecarboxamide . 2HCl. Formula: C14H21N3O . 2HCl. MW: 247.3 . 73.0. CAS Number: 129830-38-2. Purity: ≥98% (HPLC (UV)). Appearance: White to pale yellow powder. Solubility: Soluble in DMSO (25mg/ml), methanol (25mg/ml), acetonitrile (<1mg/ml), dioxan (<1mg/ml), water (very soluble). Use/Stability: Stock solutions are stable for up to 1 month when stored at -20°C. Long Term Storage: -20°C
Catalog No. 50-103-1738