Learn More
TARGETMOL CHEMICALS INC Y-27632 dihydrochloride 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T172525MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Y-27632 dihydrochloride (Y-27632 2HCl) is an orally potent, ATP-competitive inhibitor of ROCK-I and ROCK-II. Y-27632 dihydrochloride also inhibits isolation-induced apoptosis in mouse prostate stem or progenitor cells. Purity 99.67%
Catalog No. 50-287-8282