Learn More
TARGETMOL CHEMICALS INC Y-29794 Tosylate 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T3523310MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes.Y-29794 Tosylate (Y-29794) is an orally active, effective, and specific non-peptide-prolyl endopeptidase (PPCE) inhibitor. Purity 99.1%
Catalog No. 50-287-6185