Learn More
Medchemexpress LLC Y-33075 dihydrochloride | 173897-44-4 | MFCD00945328 | 98.0% | 353.25 | C16H18Cl2N4O | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1006910MM 1ML
Encompass_Preferred
Y-33075 dihydrochloride is a potent and selective Rho-associated protein kinase (ROCK) inhibitor supplied as a 10 mM solution in DMSO for laboratory research. It exhibits an IC50 of 3.6 nM against ROCK, and is characterized by a molecular weight of 353.25 and formula C16H18Cl2N4O. The compound is provided with high purity for use in biochemical and cell signaling studies.
  • Selective ROCK inhibition with IC50 of 3.6 nM.
  • Provided as a 10 mM solution in DMSO for ready-to-use assays.
  • High purity suitable for biochemical and cellular experiments.
  • Molecular weight 353.25 and molecular formula C16H18Cl2N4O.
  • Supplied in 1 mL aliquots to minimize freeze-thaw cycles.
Catalog No. 50-002-14616