Learn More
ECM Bioscience γ-Catenin Phospho-Regulation Antibody Sampler Kit, ECM Biosciences
Supplier:  ECM Bioscience CK6150
Encompass_Preferred
The γ-Catenin phospho-regulation antibody sampler kit can be used to examine phosphorylation of γ-Catenin at Tyr-133, Tyr-480, Tyr-550 and Tyr-644. The kit also includes monoclonal and polyclonal antibodies to monitor total expression levels of γ-Catenin. Applications: WB, E, ICC, IP; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8623