Learn More
Foxx Life Sciences Polypropylene Y-Type Barbed Tubing Connectors (Wye-Shaped), Three 1/4 Inch Barbs, Autoclavable, 100/PK
Supplier:  Foxx Life Sciences 2552604OEM
Encompass_Preferred
255-2604-OEM: Connector, Y Connector, Polypropylene, 1/4" Hose Barb x 1/4" Hose Barb to 1/4" Hose Barb, 100/pk. Foxx Life Sciences Y connectors are available in a variety of standard sizes. They are guaranteed to provide a secure, watertight and airtight connection. Our Y connectors are non-corrosive and durable. They are made from polypropylene.
Catalog No. 50-117-8746