Learn More
Accuflow Systems Saint-Gobain Y Connector, Polyethylene, 3/16 in, Pack of 10
Supplier:  Accuflow Systems 3072604
Encompass_Preferred
Saint-Gobain Y connectors are polyethylene barbed fittings designed for 3/16 in inner diameter tubing. The Y configuration enables splitting or combining fluid paths in low-pressure liquid and pneumatic systems. Sold as a pack of 10.
  • Polyethylene construction for general chemical compatibility and flexibility.
  • 3/16 in barbed terminations for secure tubing engagement.
  • Y configuration for splitting or merging fluid lines.
  • Pack of 10 for maintenance and spares.
  • Suitable for low-pressure liquid and pneumatic applications.
Catalog No. NC2251353