Learn More
Research Products International Corp Y Tubing Connectors, 3/16"", 12/pk
SDP
Supplier:  Research Products International Corp F19613
Encompass
Use these fittings to ensure a tight, secure connection between lengths of tubing having the same inner diameter (I.D.). Multiple barbs per arm provide additional contact between the fitting and the tubing for extra gripping power. Chemical-resistant polypropylene plastic is for use with a wide range of liquids or gases. FDA-grade allow for direct contact with food, drugs, and cosmetics. Fittings are autoclavable.
Catalog No. 50-197-8362