Learn More
Medchemexpress LLC HY-111503 5mg Medchemexpress, Y06137 CAS:2226534-49-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1115035MG
Encompass_Preferred
Medchemexpress, HY-111503 5mg Y06137 CAS:2226534-49-0 Y06137 is a potent and selective BET inhibitor for treatment of castration-resistant prostate cancer (CRPC). Y06137 binds to the BRD4(1) bromodomain with a K d of 81 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2368