Learn More
ATCC Y-1; Adrenal Cortex Tumor; Mouse (Mus musculus)
Supplier:  ATCC CCL79
Encompass
Y-1 is a cell line exhibiting epithelial morphology that was isolated from the adrenal gland of a male mouse. The mouse was tested and found negative for ectromelia virus (mousepox). This cell line was deposited by G Sato.
Catalog No. 50-238-2710