Learn More
Cayman Chemical Y27632 hydrochloride
SDP
Supplier:  Cayman Chemical 1000558350 MG
Encompass_Preferred
A potent, ATP-competitive inhibitor of ROCKs including p160ROCK (Ki = 140 nM) and ROCK2 (IC50 = 800 nM); also inhibits PRK2 with an IC50 value of 600 nM
Catalog No. 50-288-7655