Learn More
Medchemexpress LLC Yadanziolide A | 95258-14-3 | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HYN421010MM/1ML
Encompass_Preferred
Yadanziolide A, isolated from the cultivated dry seeds of *Brucea javanica*, exhibits strong antiviral activities with an IC50 of 5.5 μM against tobacco mosaic virus. It also demonstrates significant antitumor effects and is intended for research use only.
  • Isolated from *Brucea javanica*.
  • Exhibits strong antiviral activity with an IC50 of 5.5 μM against tobacco mosaic virus.
  • Demonstrates significant antitumor effects.
  • Soluble in DMSO at 100 mg/mL.
  • Powder can be stored at -20°C for 3 years or 4°C for 2 years.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-004-18541