Learn More
Medchemexpress LLC YAP/TAZ inhibitor-2 | 2762617-31-0 | 99.8% | 390.33 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY147322100MG
Encompass_Preferred
YAP/TAZ inhibitor-2 is a potent and orally active TEAD-YAP/TAZ inhibitor with an EC50 value of 3 nM. It demonstrates significant anti-proliferative and anti-tumor activity, making it a valuable compound for research in cancer biology and related fields. This inhibitor is designed to target key pathways involved in cell growth and proliferation.
  • Potent and orally active TEAD-YAP/TAZ inhibitor.
  • Exhibits anti-proliferative activity.
  • Demonstrates anti-tumor effects.
  • Valuable for cancer research.
Catalog No. 50-003-61356