Learn More
Medchemexpress LLC YAP/TAZ inhibitor-2 | 2762617-31-0 | 99.8% | 390.33 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY147322
Encompass_Preferred
YAP/TAZ inhibitor-2 is a potent and orally active TEAD-YAP/TAZ inhibitor with an EC50 value of 3 nM. It demonstrates anti-proliferative and antitumor activity.
  • Exhibits antiproliferative activity with an IC50 value of 5.33 nM in NCI-H226 cells
  • Shows anti-tumor activity in BALB/c nude mice (NCI-H226 xenograft model)
Catalog No. 50-003-63149