Learn More
Medchemexpress LLC YAP/TAZ inhibitor-2 | 2762617-31-0 | 99.8% | 390.33 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14732250MG
Encompass_Preferred
YAP/TAZ inhibitor-2 is a potent and orally active TEAD-YAP/TAZ inhibitor with an EC50 value of 3 nM. It demonstrates significant anti-proliferative and anti-tumor activity.
  • Potent and orally active TEAD-YAP/TAZ inhibitor with an EC50 value of 3 nM.
  • Shows antiproliferative activity with an IC50 value of 5.33 nM in NCI-H226 cells.
  • Exhibits anti-tumor activity in BALB/c nude mice (NCI-H226 xenograft model) without significant body weight loss.
Catalog No. 50-003-62800