Learn More
Medchemexpress LLC YAP-TEAD-IN-1 TFA | 1659305-79-9 | 99.99% | 2133.88 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP2244A10MG
Encompass_Preferred
YAP-TEAD-IN-1 TFA is a potent and competitive peptide inhibitor of YAP-TEAD interaction (IC50=25 nM). This 17mer peptide demonstrates a higher binding affinity to TEAD1 (Kd=15 nM) than YAP (50-171) (Kd= 40 nM).
  • Potent and competitive peptide inhibitor of YAP-TEAD interaction
  • IC50 of 25 nM
  • 17mer peptide
  • Higher binding affinity to TEAD1 (Kd=15 nM)
  • Active against endogenous YAP binding to GST-TEAD
  • Targets YAP/TAZ
  • Soluble in H2O at ≥ 50 mg/mL (23.43 mM)
Catalog No. 50-004-22995