Learn More
Medchemexpress LLC YAP-TEAD-IN-1 TFA | 1659305-79-9 | 99.99% | 2133.88 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP2244A5MG
Encompass_Preferred
YAP-TEAD-IN-1 TFA is a potent and competitive peptide inhibitor of YAP-TEAD interaction, demonstrating an IC50 of 25 nM. This 17mer peptide is characterized by its higher binding affinity to TEAD1 (Kd=15 nM) compared to YAP (50-171) (Kd= 40 nM). It functions by inhibiting the interaction between YAP and TEAD, key proteins involved in cell proliferation and organ size regulation.
  • Potent and competitive peptide inhibitor
  • Inhibits YAP-TEAD interaction with an IC50 of 25 nM
  • 17mer peptide structure
  • Higher binding affinity to TEAD1 (Kd=15 nM) than to YAP (50-171) (Kd= 40 nM)
  • Impacts cell proliferation and organ size regulation
Catalog No. 50-004-23616