Learn More
AdipoGen YC-1 (25 mg)
SDP
Supplier:  AdipoGen AGCR10120M025
Encompass_Preferred
Chemical. CAS: 170632-47-0. Formula: C19H16N2O2. MW: 304.4. Nitric oxide (NO) independent, superoxide-sensitive soluble guanylyl cyclase (sGC) activator. Induces concentration-dependent increase in cGMP levels. Inhibits platelet adhesion to collagen. Thombosis inhibitor. Non-specific phosphodiesterase inhibitor. Na+ channel blocker. Anticancer compound. HIF-alpha inhibitor. Blocks angiogenesis. Tumor growth inhibitor. Apoptosis inducer. NK cell differentiation enhancer. TRAIL sensitizer.
Catalog No. 50-205-3850