Learn More
Medchemexpress LLC Yeast peptone dextrose | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY172192
Encompass_Preferred
Yeast peptone dextrose is a common and nutrient-rich culture medium composed of yeast extract, peptone, and dextrose, primarily used for cultivating yeast in research settings.
  • Composed of yeast extract, peptone, and dextrose.
  • Nutrient-rich medium.
  • Primarily for cultivating yeast.
  • For research use only.
Catalog No. 50-003-84194