Learn More
Biotium Yeast Viability Apoptosis and Viability Assays Dead cell stains Live or Dye 850 870 1/EA
Supplier:  Biotium 32022
Encompass_Preferred
Live-or-Dye Fixable Viability Staining Kits are designed for discrimination between live and dead cells during flow cytometry Live-or-Dye 850/870 has emission at 870 nm and can be excited by an 808 laser line 200 assays per kit
Catalog No. 50-005-01485