Learn More
ATCC Yellow fever virus, 17D vaccine strain (attenuated) - frozen viral preparation, BSL-2 (ATCC VR-1506)
Supplier: ATCC VR1506
An attenuated Yellow fever virus (17D) vaccine strain supplied as a frozen viral preparation for research and vaccine development. The strain was derived from the Asibi isolate and adapted for propagation in Vero cells; handle under BSL-2 containment.
- Supplied as a frozen viral preparation.
- Store at -70°C or colder.
- Attenuated 17D vaccine strain derived from the Asibi isolate.
- Adapted for propagation in Vero cells (CCL-81).
- Intended for vaccine development and production research.
- Biosafety level 2 containment recommended.
- Geographical origin: Ghana.