Learn More
ATCC Yersinia enterocolitica; subsp. enterocolitica
Supplier:  ATCC 23715
Encompass
Yersinia enterocolitica subsp. enterocolitica strain Billups-1803-68 is a whole-genome sequenced bacterial strain that was isolated in 1968 from petechiae at the anterior chamber of a human eye. This strain has applications in enteric disease research, infectious disease research, and media testing.
Catalog No. 50-238-2990