Learn More
ATCC Prevotella pallens
Supplier:  ATCC 700821
Encompass
Prevotella pallens strain NCTC 13042 is a bacterial type strain that was isolated from the saliva of a human child. This strain is propagated anaerobically on modified chopped meat medium and trypticase soy medium.
Catalog No. 50-238-1118