Learn More
Sino Biological Yellow fever virus (YFV) (strain 17D vaccine) Envelope Protein (His Tag)
Supplier:  Sino Biological 41052V08B100
Encompass_Preferred
A DNA sequence encoding the Yellow fever virus (YFV) (strain 17D vaccine) Envelope Protein (P03314) (Ala286-Ser680) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-3328