Learn More
Medchemexpress LLC YH-53 | 1471484-62-4 | 98.8% | 575.68 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13931150MG
Encompass_Preferred
YH-53 is a potent peptidomimetic 3CLpro inhibitor featuring a unique benzothiazolyl ketone structure. It exhibits strong inhibitory activity against SARS-CoV-1 3CLpro (Ki: 6.3 nM) and SARS-CoV-2 3CLpro (Ki: 34.7 nM), effectively blocking SARS-CoV-2 replication. This compound holds potential for COVID-19 research.
  • Potent 3CLpro inhibitor
  • Blocks SARS-CoV-2 replication
  • Peptidomimetic compound with unique benzothiazolyl ketone
  • Potential for COVID-19 research
  • Low cytotoxicity in Vero cells (CC50 > 100 μM)
Catalog No. 50-003-51337