Learn More
TARGETMOL CHEMICALS INC YHO-13351 FREE BASE 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1336510MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. YHO-13351 free base is the water-soluble prodrug of YHO-13177 which is an inhibitor of BCRP . purity: 100%
Catalog No. 50-257-7395