Learn More
Medchemexpress LLC YJ1206 | 3053716-98-3 | 96.9% | 894.01 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY168555
Encompass_Preferred
YJ1206 is an orally active, selective CDK12/CDK13 PROTAC degrader. It increases DNA damage, induces apoptosis, and promotes tumor regression in orthotopic WA74 patient-derived xenograft (PDX) mice models of resistant prostate cancer. It suppresses tumor growth in vivo in conjunction with AKT pathway inhibitors.
  • Increases DNA damage and induces apoptosis.
  • Promotes tumor regression in models of resistant prostate cancer.
  • Suppresses tumor growth when combined with AKT pathway inhibitors.
  • Composed of a CDK12/CDK13 degradation agent, a linker, and a VHL E3 ubiquitin ligase.
Catalog No. 50-003-93009