Learn More
Medchemexpress LLC YM-1 | 409086-68-6 | 98.49% | 417.98 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY18399100MG
Encompass_Preferred
YM-1 is a stable MKT-077 analog and an orally active Hsp70 inhibitor. It induces cell death in HeLa cells and up-regulates the levels of p53 and p21 proteins. For research use only, not for sale to patients.
  • Stable MKT-077 analog
  • Orally active Hsp70 inhibitor
  • Induces cell death of HeLa cells
  • Up-regulates p53 and p21 proteins
  • Promotes Hsp70-dependent steps in nNOS maturation
  • Partially blocks formation of the ATP-bound form of Hsp70
  • Activates binding of Hsp70 to its unfolded substrate
  • Converts Hsp70 to its tight-affinity conformation with an IC50 of 8.2 μM
  • Promotes nNOS ubiquitination
  • Weakens dihydrotestosterone (DHT)-dependent eye degeneration phenotype and rescues DHT-dependent pupal toxicity of polyQ AR in Drosophila
Catalog No. 50-003-88083