Learn More
Medchemexpress LLC YM-244769 | 838819-70-8 | MFCD28160863 | 99.2% | 443.47 g·mol⁻¹ | C26H22FN3O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY136182A5MG
Encompass_Preferred
YM-244769 is a potent, selective, orally active inhibitor of the sodium-calcium exchanger (NCX) with preferential activity toward the NCX3 isoform. It suppresses NCX-mediated Ca2+ entry and has been used in cellular and in vivo studies demonstrating neuroprotective effects and effects on urine volume and electrolyte excretion in mice.
  • Preferentially inhibits NCX3 with IC50 ≈ 18 nM.
  • Also active against NCX1 and NCX2 (IC50 68 nM and 96 nM).
  • Orally active and shows neuroprotective effects in hypoxia/reoxygenation models.
  • High purity (≈99.2%) and solid, off-white to light yellow appearance.
  • Soluble in DMSO (≈120 mg/mL); requires ultrasonic assistance.
  • Store sealed at 4°C; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-22109