Learn More
Apexbio Technology LLC YM155 781661-94-7 25mg
SDP
Supplier:  Apexbio Technology LLC A422125
Encompass_Preferred
YM155 (Sepantronium Bromide CAS 781661-94-7) is a small-molecule inhibitor targeting survivin the smallest protein member of the inhibitor of apoptosis (IAP) gene family YM155 potently suppresses survivin expression while demonstrating minimal influence on other IAP family members or BCL-2-associated proteins Preclinical research shows YM155 inhibits proliferation and induces apoptosis across various human cancer cell lines including non-small cell lung cancer (NSCLC) melanoma bladder cancer aggressive non-Hodgkin lymphoma triple-negative breast cancer (TNBC) and Wilms tumor models In animal xenograft studies YM155 administration results in tumor regression reduces spontaneous metastasis and notably prolongs survival
Catalog No. 50-000-62530