Learn More
Medchemexpress LLC YM511 5mg | 148869-05-0 | 5MG
SDP
Supplier:  Medchemexpress LLC HY1051845MG
Encompass_Preferred
YM511 is a highly specific non-steroidal aromatase inhibitor YM511 inhibits aromatase activities in microsomes from rat ovary and human placenta competitively (IC50s of 0 4 and 0 12 nM respectively) YM511 slightly inhibits production of other steroid hormones YM511 has the potential for suppressing estrogen-dependent action research without affecting serum levels of other steroid hormones[1
Catalog No. 50-002-44098