Learn More
YMC America, Inc YMC-Triart Prep C18-S column, 120 Å, 10 µm, 150mm L, 10mm ID
SDP
Supplier:  YMC America, Inc TAS12S111510WT
Encompass_Preferred
YMC-Triart C18 is an organic/inorganic hybrid silica particle derivatized with C18 (L1 USP designation). It is manufactured using state-of-the-art technologies that define materials, granulation, and surface modification. This results in a product that exhibits superior peak shape and chemical durability, even at high temperatures and wide pH ranges.YMC-Triart C18 is designed to retain both moderate hydrogen bonding capacity and hydrophobicity on the surface by optimizing the bonding density of the C18 phase. This allows it to be used for the analysis of polar compounds under 100% aqueous mobile phase conditions.
Catalog No. NC3802126