Learn More
YMC America, Inc YMCbasic 5um 150x0.3mm 1/16
SDP
Supplier:  YMC America, Inc BA99S0515H0AU
Encompass
YMCbasic is a unique reversed-phase column designed for analysis of pharmaceuticals and basic compounds Innovative silica synthesis and state-of-the-art silane bonding result in a chromatographic material with neutral silanols This minimizes interactions with both basic and acidic compounds
Catalog No. 50-008-79526